2-Bromo-4-(trifluoromethoxy)anisole 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-1-methoxy-4-(trifluoromethoxy)benzene
PC3124-1G
PC3124-5G
PC3124-25G
PC3124-100G
APOSPC3124-1GEA
2480
CZK
APOSPC3124-1G
APOSPC3124-5G
APOSPC3124-25G
APOSPC3124-100G
2-Bromo-4-(trifluoromethoxy)anisole 97%
2-Bromo-4-(trifluoromethoxy)anisole
Vzorec:
C₈H₆BrF₃O₂ MW: 271,031 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973759 CAS číslo: 200956-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...