2',5'-Bis(2,2,2-trifluoroethoxy)acetophenone 95%
Dodavatel: Apollo Scientific
PC3053-1G
PC3053-250MG
PC3053-5G
APOSPC3053-1GEA
11680
CZK
APOSPC3053-1G
APOSPC3053-250MG
APOSPC3053-5G
2',5'-Bis(2,2,2-trifluoroethoxy)acetophenone 95%
2',5'-Bis(2,2,2-trifluoroethoxy)acetophenone
Vzorec:
C₁₂H₁₀F₆O₃ MW: 316,196 g/mol Bod tání: 90…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042271 CAS číslo: 76784-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...