Tisk…

3-Bromo-4-methyl-5-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific

PC303608-1G PC303608-5G
APOSPC303608-1GEA 10910 CZK
APOSPC303608-1G APOSPC303608-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzonitrile
3-Bromo-4-methyl-5-(trifluoromethyl)benzonitrile
Vzorec: C₉H₅BrF₃N
MW: 264,042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640181
CAS číslo: 2383004-77-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR