3-Bromo-4-methyl-5-(trifluoromethyl)benzoyl chloride
Dodavatel: Apollo Scientific
PC303606-1G
PC303606-5G
APOSPC303606-1GEA
8930
CZK
APOSPC303606-1G
APOSPC303606-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzoyl chloride
3-Bromo-4-methyl-5-(trifluoromethyl)benzoyl chloride
Vzorec:
C₉H₅BrClF₃O MW: 301,488 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640179 CAS číslo: 2383663-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...