3-Bromo-4-methyl-5-(trifluoromethyl)benzylamine
Dodavatel: Apollo Scientific
PC303604-1G
PC303604-5G
APOSPC303604-1GEA
12240
CZK
APOSPC303604-1G
APOSPC303604-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzylamine
3-Bromo-4-methyl-5-(trifluoromethyl)benzylamine
Vzorec:
C₉H₉BrF₃N MW: 268.074 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640177 CAS číslo: 2383901-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...