Tisk…

3-Bromo-2-chloro-5-(trifluoromethyl)phenylacetic acid

Dodavatel: Apollo Scientific

PC303579-1G PC303579-5G
APOSPC303579-1GEA 11910 CZK
APOSPC303579-1G APOSPC303579-5G
3-Bromo-2-chloro-5-(trifluoromethyl)phenylacetic acid
3-Bromo-2-chloro-5-(trifluoromethyl)phenylacetic acid
Vzorec: C₉H₅BrClF₃O₂
MW: 317,487 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640156
CAS číslo: 2090523-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR