Tisk…

3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol

Dodavatel: Apollo Scientific

PC303576-1G PC303576-5G
APOSPC303576-1GEA 6580 CZK
APOSPC303576-1G APOSPC303576-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₅BrClF₃O
MW: 289.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640153
CAS číslo: 2092609-70-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR