Tisk…

3-Bromo-2-chloro-5-(trifluoromethyl)benzaldehyde 98%

Dodavatel: Apollo Scientific

PC303575-1G PC303575-5G
APOSPC303575-1GEA 6220 CZK
APOSPC303575-1G APOSPC303575-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzaldehyde 98%
3-Bromo-2-chloro-5-(trifluoromethyl)benzaldehyde
Vzorec: C₈H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD32640152
CAS číslo: 2090464-41-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR