Tisk…

3-Bromo-4-chloro-5-(trifluoromethyl)benzoyl chloride

Dodavatel: Apollo Scientific

PC303570-1G PC303570-5G
APOSPC303570-1GEA 6580 CZK
APOSPC303570-1G APOSPC303570-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzoyl chloride
3-Bromo-4-chloro-5-(trifluoromethyl)benzoyl chloride
Vzorec: C₈H₂BrCl₂F₃O
MW: 321,906 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640148
CAS číslo: 2385595-04-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR