3-Bromo-4-chloro-5-(trifluoromethyl)benzyl alcohol
Dodavatel: Apollo Scientific
PC303564-1G
PC303564-5G
APOSPC303564-1GEA
6580
CZK
APOSPC303564-1G
APOSPC303564-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl alcohol
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl alcohol
Vzorec:
C₈H₅BrClF₃O MW: 289.477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640142 CAS číslo: 2384801-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...