2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile
Dodavatel: Apollo Scientific
PC303562-1G
PC303562-5G
APOSPC303562-1GEA
9590
CZK
APOSPC303562-1G
APOSPC303562-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile
Vzorec:
C₉H₄BrF₄N MW: 282,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640140 CAS číslo: 2091737-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...