2-Bromo-5-fluoro-4-(trifluoromethyl)benzoic acid 97%
Dodavatel: Apollo Scientific
PC303557-1G
PC303557-5G
APOSPC303557-1GEA
6550
CZK
APOSPC303557-1G
APOSPC303557-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzoic acid 97%
2-Bromo-5-fluoro-4-(trifluoromethyl)benzoic acid
Vzorec:
C₈H₃BrF₄O₂ MW: 287,006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640135 CAS číslo: 2090964-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...