Tisk…

2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl alcohol

Dodavatel: Apollo Scientific

PC303553-1G PC303553-5G
APOSPC303553-1GEA 0 CZK
APOSPC303553-1G APOSPC303553-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl alcohol
2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₅BrF₄O
MW: 273,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640131
CAS číslo: 2090282-92-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR