5-Bromo-2-fluoro-3-(trifluoromethyl)benzylamine
Dodavatel: Apollo Scientific
PC303545-1G
PC303545-5G
APOSPC303545-1GEA
12240
CZK
APOSPC303545-1G
APOSPC303545-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzylamine
5-Bromo-2-fluoro-3-(trifluoromethyl)benzylamine
Vzorec:
C₈H₆BrF₄N MW: 272.038 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640126 CAS číslo: 2386111-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...