3-Bromo-6-fluoro-2-(trifluoromethyl)phenylacetonitrile 97%
Dodavatel: Apollo Scientific
PC303537-1G
PC303537-5G
APOSPC303537-1GEA
9260
CZK
APOSPC303537-1G
APOSPC303537-5G
3-Bromo-6-fluoro-2-(trifluoromethyl)phenylacetonitrile 97%
3-Bromo-6-fluoro-2-(trifluoromethyl)phenylacetonitrile
Vzorec:
C₉H₄BrF₄N MW: 282,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640119 CAS číslo: 2091737-61-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...