Tisk…

2-Bromo-4,6-bis(trifluoromethyl)benzylamine

Dodavatel: Apollo Scientific

PC303533-1G PC303533-5G
APOSPC303533-1GEA 12240 CZK
APOSPC303533-1G APOSPC303533-5G
2-Bromo-4,6-bis(trifluoromethyl)benzylamine
2-Bromo-4,6-bis(trifluoromethyl)benzylamine
Vzorec: C₉H₆BrF₆N
MW: 322,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28727989
CAS číslo: 1805512-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR