Tisk…

2-Bromo-4,6-bis(trifluoromethyl)benzyl alcohol

Dodavatel: Apollo Scientific

PC303529-1G PC303529-5G
APOSPC303529-1GEA 8930 CZK
APOSPC303529-1G APOSPC303529-5G
2-Bromo-4,6-bis(trifluoromethyl)benzyl alcohol
2-Bromo-4,6-bis(trifluoromethyl)benzyl alcohol
Vzorec: C₉H₅BrF₆O
MW: 323.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28727974
CAS číslo: 1805512-50-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR