Tisk…

3-Bromo-4-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC303525-1G PC303525-5G
APOSPC303525-1GEA 9260 CZK
APOSPC303525-1G APOSPC303525-5G
3-Bromo-4-(trifluoromethoxy)phenylacetonitrile 97%
3-Bromo-4-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₉H₅BrF₃NO
MW: 280.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640114
CAS číslo: 2167710-86-1
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR