3-Bromo-5,6-difluoro-2-methoxyphenylacetic acid
Dodavatel: Apollo Scientific
PC303491-1G
PC303491-5G
APOSPC303491-1GEA
11910
CZK
APOSPC303491-1G
APOSPC303491-5G
3-Bromo-5,6-difluoro-2-methoxyphenylacetic acid
3-Bromo-5,6-difluoro-2-methoxyphenylacetic acid
Vzorec:
C₉H₇BrF₂O₃ MW: 281,051 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640085 CAS číslo: 1889215-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...