5-Bromo-4-chloro-2-fluorobenzyl alcohol 98%
Dodavatel: Apollo Scientific
Synonyms:
(5-Bromo-4-chloro-2-fluorophenyl)methanol
PC303477-1G
PC303477-5G
APOSPC303477-1GEA
6580
CZK
APOSPC303477-1G
APOSPC303477-5G
5-Bromo-4-chloro-2-fluorobenzyl alcohol 98%
5-Bromo-4-chloro-2-fluorobenzyl alcohol
Vzorec:
C₇H₅BrClFO MW: 239,469 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28060809 CAS číslo: 1785275-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...