5-Bromo-3-chloro-2-fluorobenzylamine
Dodavatel: Apollo Scientific
PC303463-1G
PC303463-250MG
APOSPC303463-1GEA
19010
CZK
APOSPC303463-1G
APOSPC303463-250MG
5-Bromo-3-chloro-2-fluorobenzylamine
5-Bromo-3-chloro-2-fluorobenzylamine
Vzorec:
C₇H₆BrClFN MW: 238,485 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23163937 CAS číslo: 1386459-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...