5-Bromo-3-chloro-2-fluorobenzonitrile
Dodavatel: Apollo Scientific
PC303460-1G
PC303460-5G
PC303460-250MG
APOSPC303460-1GEA
8730
CZK
APOSPC303460-1G
APOSPC303460-5G
APOSPC303460-250MG
5-Bromo-3-chloro-2-fluorobenzonitrile
5-Bromo-3-chloro-2-fluorobenzonitrile
Vzorec:
C₇H₂BrClFN Storage Temperature: Ambient |
MDL Number:
MFCD09800811 CAS číslo: 1000577-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...