5-Bromo-3-chloro-2-fluorobenzaldehyde 97%
Dodavatel: Apollo Scientific
PC303452-1G
PC303452-5G
APOSPC303452-1GEA
1210
CZK
APOSPC303452-1G
APOSPC303452-5G
5-Bromo-3-chloro-2-fluorobenzaldehyde 97%
5-Bromo-3-chloro-2-fluorobenzaldehyde
Vzorec:
C₇H₃BrClFO MW: 237,454 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18783162 CAS číslo: 1280786-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...