3-Bromo-2,6-difluorophenylacetic acid
Dodavatel: Apollo Scientific
PC303448-1G
APOSPC303448-1GEA
23970
CZK
APOSPC303448-1G
3-Bromo-2,6-difluorophenylacetic acid
3-Bromo-2,6-difluorophenylacetic acid
Vzorec:
C₈H₅BrF₂O₂ MW: 251,025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16987590 CAS číslo: 1250443-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...