Tisk…

3-Bromo-2,6-difluorobenzyl alcohol

Dodavatel: Apollo Scientific

Synonyms: (3-Bromo-2,6-difluorophenyl)methanol

PC303446-1G PC303446-5G
APOSPC303446-1GEA 2650 CZK
APOSPC303446-1G APOSPC303446-5G
3-Bromo-2,6-difluorobenzyl alcohol
3-Bromo-2,6-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MW: 223,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483917
CAS číslo: 438050-05-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR