Tisk…

2,3,4-Trifluoro-5-methylaniline

Dodavatel: Apollo Scientific

PC303443-1G PC303443-5G
APOSPC303443-1GEA 9920 CZK
APOSPC303443-1G APOSPC303443-5G
2,3,4-Trifluoro-5-methylaniline
2,3,4-Trifluoro-5-methylaniline
Vzorec: C₇H₆F₃N
MW: 161,124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687840
CAS číslo: 1536760-94-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR