Tisk…

2,3,4-Trifluoro-5-methylbenzoic acid 97%

Dodavatel: Apollo Scientific

PC303439-1G PC303439-5G
APOSPC303439-1GEA 4960 CZK
APOSPC303439-1G APOSPC303439-5G
2,3,4-Trifluoro-5-methylbenzoic acid 97%
2,3,4-Trifluoro-5-methylbenzoic acid
Vzorec: C₈H₅F₃O₂
MW: 190,119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20643360
CAS číslo: 65829-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR