Tisk…

5-Bromo-2,3-difluorobenzamide

Dodavatel: Apollo Scientific

PC303430-1G PC303430-5G
APOSPC303430-1GEA 6950 CZK
APOSPC303430-1G APOSPC303430-5G
5-Bromo-2,3-difluorobenzamide
5-Bromo-2,3-difluorobenzamide
Vzorec: C₇H₄BrF₂NO
MW: 236,014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737879
CAS číslo: 1626337-90-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR