Tisk…

5-Bromo-2,3-difluorobenzylamine

Dodavatel: Apollo Scientific

PC303427-1G PC303427-5G
APOSPC303427-1GEA 10580 CZK
APOSPC303427-1G APOSPC303427-5G
5-Bromo-2,3-difluorobenzylamine
5-Bromo-2,3-difluorobenzylamine
Vzorec: C₇H₆BrF₂N
MW: 222.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972476
CAS číslo: 887585-92-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR