Tisk…

3,5-Dimethyl-2,4,6-trifluorobenzaldehyde 97%

Dodavatel: Apollo Scientific

PC303424-1G PC303424-5G
APOSPC303424-1GEA 8270 CZK
APOSPC303424-1G APOSPC303424-5G
3,5-Dimethyl-2,4,6-trifluorobenzaldehyde 97%
3,5-Dimethyl-2,4,6-trifluorobenzaldehyde
Vzorec: C₉H₇F₃O
MW: 188,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29764385
CAS číslo: 1951441-62-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR