4-(4-Chloro-3-(trifluoromethyl)phenoxy)aniline 97%
Dodavatel: Apollo Scientific
PC303410-1G
PC303410-5G
APOSPC303410-1GEA
2070
CZK
APOSPC303410-1G
APOSPC303410-5G
4-(4-Chloro-3-(trifluoromethyl)phenoxy)aniline 97%
4-(4-Chloro-3-(trifluoromethyl)phenoxy)aniline
Vzorec:
C₁₃H₉ClF₃NO MW: 287,665 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28404281 CAS číslo: 1369254-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...