Tisk…

6-Chloro-3-fluoro-2-methoxybenzaldehyde 97%

Dodavatel: Apollo Scientific

PC303409-1G PC303409-250MG
APOSPC303409-1GEA 18520 CZK
APOSPC303409-1G APOSPC303409-250MG
6-Chloro-3-fluoro-2-methoxybenzaldehyde 97%
6-Chloro-3-fluoro-2-methoxybenzaldehyde
Vzorec: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24687551
CAS číslo: 1350325-23-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR