Tisk…

6-Chloro-2,4-difluoro-3-methylbenzonitrile 97%

Dodavatel: Apollo Scientific

PC303406-1G PC303406-250MG
APOSPC303406-1GEA 27780 CZK
APOSPC303406-1G APOSPC303406-250MG
6-Chloro-2,4-difluoro-3-methylbenzonitrile 97%
6-Chloro-2,4-difluoro-3-methylbenzonitrile
Vzorec: C₈H₄ClF₂N
MW: 187,574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640060
CAS číslo: 2415751-66-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR