Tisk…

3-Fluoro-2,4-dimethylbenzonitrile 97%

Dodavatel: Apollo Scientific

PC303400-1G PC303400-5G
APOSPC303400-1GEA 2200 CZK
APOSPC303400-1G APOSPC303400-5G
3-Fluoro-2,4-dimethylbenzonitrile 97%
3-Fluoro-2,4-dimethylbenzonitrile
Vzorec: C₉H₈FN
MW: 149,165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794310
CAS číslo: 1806330-91-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR