3-Fluoro-2,4-dimethylbenzyl bromide 97%
Dodavatel: Apollo Scientific
PC303399-1G
PC303399-5G
APOSPC303399-1GEA
4860
CZK
APOSPC303399-1G
APOSPC303399-5G
3-Fluoro-2,4-dimethylbenzyl bromide 97%
3-Fluoro-2,4-dimethylbenzyl bromide
Vzorec:
C₉H₁₀BrF Storage Temperature: Ambient |
MDL Number:
MFCD28794354 CAS číslo: 1806352-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...