Tisk…

4-Bromo-2-chloro-5-(trifluoromethoxy)phenol 98%

Dodavatel: Apollo Scientific

PC303391-1G PC303391-250MG
APOSPC303391-1GEA 19410 CZK
APOSPC303391-1G APOSPC303391-250MG
4-Bromo-2-chloro-5-(trifluoromethoxy)phenol 98%
4-Bromo-2-chloro-5-(trifluoromethoxy)phenol
Vzorec: C₇H₃BrClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD22419064
CAS číslo: 1824642-82-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR