Tisk…

2-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid 98%

Dodavatel: Apollo Scientific

PC303379-1G PC303379-250MG
APOSPC303379-1GEA 26420 CZK
APOSPC303379-1G APOSPC303379-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₈F₄O₂
MW: 236,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640050
CAS číslo: 2383659-72-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR