2-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol 98%
Dodavatel: Apollo Scientific
PC303377-1G
PC303377-250MG
APOSPC303377-1GEA
16140
CZK
APOSPC303377-1G
APOSPC303377-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol 98%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzyl alcohol
Vzorec:
C₉H₈F₄O MW: 208,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31916392 CAS číslo: 2092563-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...