4-Bromo-2,5-bis(trifluoromethyl)benzoic acid 98%
Dodavatel: Apollo Scientific
PC303366-1G
PC303366-250MG
APOSPC303366-1GEA
14880
CZK
APOSPC303366-1G
APOSPC303366-250MG
4-Bromo-2,5-bis(trifluoromethyl)benzoic acid 98%
4-Bromo-2,5-bis(trifluoromethyl)benzoic acid
Vzorec:
C₉H₃BrF₆O₂ MW: 337.013 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28727899 CAS číslo: 1805511-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...