2-Chloro-3-fluoro-6-methoxybenzaldehyde 97%
Dodavatel: Apollo Scientific
PC303365-1G
PC303365-250MG
APOSPC303365-1GEA
12630
CZK
APOSPC303365-1G
APOSPC303365-250MG
2-Chloro-3-fluoro-6-methoxybenzaldehyde 97%
2-Chloro-3-fluoro-6-methoxybenzaldehyde
Vzorec:
C₈H₆ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18432476 CAS číslo: 1263378-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...