Tisk…

5-Chloro-2,4-difluorobenzyl bromide 97%

Dodavatel: Apollo Scientific

PC303355-5G PC303355-1G
APOSPC303355-5GEA 23810 CZK
APOSPC303355-5G APOSPC303355-1G
5-Chloro-2,4-difluorobenzyl bromide 97%
5-Chloro-2,4-difluorobenzyl bromide
Vzorec: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD27932574
CAS číslo: 915409-64-2
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR