5-Chloro-2,4-difluorobenzaldehyde 98%
Dodavatel: Apollo Scientific
PC303353-5G
PC303353-1G
PC303353-10G
APOSPC303353-5GEA
4270
CZK
APOSPC303353-5G
APOSPC303353-1G
APOSPC303353-10G
5-Chloro-2,4-difluorobenzaldehyde 98%
5-Chloro-2,4-difluorobenzaldehyde
Vzorec:
C₇H₃ClF₂O Storage Temperature: Ambient |
MDL Number:
MFCD07782052 CAS číslo: 695187-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...