Tisk…

2-Chloro-4-fluoro-5-methylbenzylamine

Dodavatel: Apollo Scientific

PC303348-1G PC303348-5G
APOSPC303348-1GEA 9660 CZK
APOSPC303348-1G APOSPC303348-5G
2-Chloro-4-fluoro-5-methylbenzylamine
2-Chloro-4-fluoro-5-methylbenzylamine
Vzorec: C₈H₉ClFN
MW: 173,615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602108
CAS číslo: 1365968-65-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR