Tisk…

2-Chloro-4-fluoro-5-methylbenzamide 97%

Dodavatel: Apollo Scientific

PC303344-5G PC303344-1G
APOSPC303344-5GEA 24340 CZK
APOSPC303344-5G APOSPC303344-1G
2-Chloro-4-fluoro-5-methylbenzamide 97%
2-Chloro-4-fluoro-5-methylbenzamide
Vzorec: C₈H₇ClFNO
MW: 187,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751763
CAS číslo: 1805524-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR