Tisk…

2-Chloro-4-fluoro-5-methylbenzoic acid 97%

Dodavatel: Apollo Scientific
Varování

PC303342-5G PC303342-1G
APOSPC303342-5GEA 11870 CZK
APOSPC303342-5G APOSPC303342-1G
2-Chloro-4-fluoro-5-methylbenzoic acid 97%
2-Chloro-4-fluoro-5-methylbenzoic acid
Vzorec: C₈H₆ClFO₂
MW: 188,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20658460
CAS číslo: 208165-96-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR