2-Chloro-4-fluoro-5-methylbenzaldehyde 98%
Dodavatel: Apollo Scientific
PC303337-1G
PC303337-250MG
APOSPC303337-1GEA
24340
CZK
APOSPC303337-1G
APOSPC303337-250MG
2-Chloro-4-fluoro-5-methylbenzaldehyde 98%
2-Chloro-4-fluoro-5-methylbenzaldehyde
Vzorec:
C₈H₆ClFO MW: 172,584 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24034192 CAS číslo: 1503624-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...