Tisk…

4-Chloro-2-fluoro-5-methylbenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

PC303336-1G PC303336-250MG PC303336-5G
APOSPC303336-1GEA 17990 CZK
APOSPC303336-1G APOSPC303336-250MG APOSPC303336-5G
4-Chloro-2-fluoro-5-methylbenzonitrile
4-Chloro-2-fluoro-5-methylbenzonitrile
Vzorec: C₈H₅ClFN
MW: 169.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658721
CAS číslo: 1126424-34-7
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR