α,2-Dimethyl-3-(trifluoromethyl)benzyl bromide 98%
Dodavatel: Apollo Scientific
PC303332-5G
PC303332-1G
APOSPC303332-5GEA
27250
CZK
APOSPC303332-5G
APOSPC303332-1G
α,2-Dimethyl-3-(trifluoromethyl)benzyl bromide 98%
α,2-Dimethyl-3-(trifluoromethyl)benzyl bromide
Vzorec:
C₁₀H₁₀BrF₃ MW: 267,086 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30502875 CAS číslo: 2149597-33-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...