Tisk…

4-Chloro-2-fluoro-5-methylbenzylamine

Dodavatel: Apollo Scientific

PC303331-1G PC303331-5G
APOSPC303331-1GEA 9000 CZK
APOSPC303331-1G APOSPC303331-5G
4-Chloro-2-fluoro-5-methylbenzylamine
4-Chloro-2-fluoro-5-methylbenzylamine
Vzorec: C₈H₉ClFN
Storage Temperature: Ambient
MDL Number: MFCD23712148
CAS číslo: 1427373-84-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR