Tisk…

4-Chloro-2-fluoro-5-methylbenzamide 98%

Dodavatel: Apollo Scientific

PC303327-5G PC303327-1G
APOSPC303327-5GEA 22780 CZK
APOSPC303327-5G APOSPC303327-1G
4-Chloro-2-fluoro-5-methylbenzamide 98%
4-Chloro-2-fluoro-5-methylbenzamide
Vzorec: C₈H₇ClFNO
MW: 187,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751776
CAS číslo: 1807267-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR